Product Details

SNP ID
hCV29904584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133255868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCCGTTGGCCTGGTCGACCATC[C/A]TGGCCTGGTGGCAGGCCCTGGTGAG
Phenotype
MIM: 110300
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
ABO PubMed Links

Gene Details

Gene
ABO
Gene Name
ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020469.2 888 Missense Mutation AGG,ATG R288M NP_065202.2

View Full Product Details