Product Details

SNP ID
rs9925515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14309122 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGCACTGAGACCTATTTAGCCA[A/G]TTATTGAGAAGATGAGGTCCCTTTC
Phenotype
MIM: 614734 MIM: 614735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR193B PubMed Links

Gene Details

Gene
MIR193B
Gene Name
microRNA 193b
There are no transcripts associated with this gene.

Gene
MIR193BHG
Gene Name
MIR193B host gene
There are no transcripts associated with this gene.

Gene
MIR365A
Gene Name
microRNA 365a
There are no transcripts associated with this gene.

View Full Product Details