Product Details
- SNP ID
-
rs10511910
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:33029042 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAGCAGGTGACTTCATACTTCTAA[C/T]CTCAGTGTCCTTATGAGATCTTGAC
- Phenotype
-
MIM: 606350
MIM: 602837
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APTX
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76432063] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APTX
- Gene Name
- aprataxin
There are no transcripts associated with this gene.
- Gene
- DNAJA1
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A1
View Full Product Details