Product Details

SNP ID
rs6052095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3822799 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTGTGCCTGTAGACTTTATGAAT[C/T]TCTAACGAAGAGAGAGGATGTGGTA
Phenotype
MIM: 614824
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP5S1 PubMed Links

Gene Details

Gene
AP5S1
Gene Name
adaptor related protein complex 5 sigma 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204446.1 Intron NP_001191375.1
NM_001204447.1 Intron NP_001191376.1
NM_018347.2 Intron NP_060817.1

View Full Product Details