Product Details

SNP ID
rs10177845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27901304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTAAGTGTCTGTTCAGCATTCT[A/G]GAATGATGCCTGGCAAATACAACAG
Phenotype
MIM: 610497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRE PubMed Links

Gene Details

Gene
BRE
Gene Name
brain and reproductive organ-expressed (TNFRSF1A modulator)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261840.1 Intron NP_001248769.1
NM_004899.4 Intron NP_004890.2
NM_199191.2 Intron NP_954661.1
NM_199192.2 Intron NP_954662.1
NM_199193.2 Intron NP_954663.1
NM_199194.2 Intron NP_954664.1

View Full Product Details