Product Details

SNP ID
rs7184225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89877922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATAAAACTTATCTAAATTGACA[C/T]GTGAAGAAATAAAAAAATCTGAGCC
Phenotype
MIM: 609217 MIM: 612326
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPIRE2 PubMed Links

Gene Details

Gene
SPIRE2
Gene Name
spire type actin nucleation factor 2
There are no transcripts associated with this gene.

Gene
TCF25
Gene Name
transcription factor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014972.2 Intron NP_055787.1
XM_005256297.2 Intron XP_005256354.1
XM_005256298.2 Intron XP_005256355.1
XM_005256299.2 Intron XP_005256356.1
XM_011522956.2 Intron XP_011521258.1
XM_011522957.2 Intron XP_011521259.1
XM_011522958.2 Intron XP_011521260.1
XM_011522959.2 Intron XP_011521261.1
XM_011522960.2 Intron XP_011521262.1
XM_011522961.2 Intron XP_011521263.1
XM_017023047.1 Intron XP_016878536.1
XM_017023048.1 Intron XP_016878537.1
XM_017023049.1 Intron XP_016878538.1
XM_017023050.1 Intron XP_016878539.1
XM_017023051.1 Intron XP_016878540.1
XM_017023052.1 Intron XP_016878541.1
XM_017023053.1 Intron XP_016878542.1
XM_017023054.1 Intron XP_016878543.1
XM_017023055.1 Intron XP_016878544.1
XM_017023056.1 Intron XP_016878545.1
XM_017023057.1 Intron XP_016878546.1
XM_017023058.1 Intron XP_016878547.1

View Full Product Details