Product Details

SNP ID
rs9837396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49238428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGATTTTATTAAGGTGTGCACC[A/G]GCTCAGCGGATTCGCATCCAAAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC36 PubMed Links

Gene Details

Gene
CCDC36
Gene Name
coiled-coil domain containing 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135197.1 Intron NP_001128669.1
NM_178173.3 Intron NP_835467.2
XM_006713124.3 Intron XP_006713187.1
XM_011533670.2 Intron XP_011531972.1
XM_011533671.2 Intron XP_011531973.1
XM_011533672.2 Intron XP_011531974.1
XM_011533673.2 Intron XP_011531975.1

View Full Product Details