Product Details

SNP ID
rs4794854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29397985 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAGCAGCCTCTACCTTCAGGGCC[C/G]AAGCCGTGAGTAGCTGAGACTGCAG
Phenotype
MIM: 610266
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371931 PubMed Links

Gene Details

Gene
LOC105371931
Gene Name
uncharacterized LOC105371931
There are no transcripts associated with this gene.

Gene
MIR4523
Gene Name
microRNA 4523
There are no transcripts associated with this gene.

Gene
TAOK1
Gene Name
TAO kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020791.2 Intron NP_065842.1
NM_025142.1 Intron NP_079418.1
XM_011525060.2 Intron XP_011523362.1

View Full Product Details