Product Details

SNP ID
rs9916882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4165877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCACTTCAATTCATATCCGAA[C/T]CATGATTCTCTTACTTTCTAACTTA
Phenotype
MIM: 607927
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKFY1 PubMed Links

Gene Details

Gene
ANKFY1
Gene Name
ankyrin repeat and FYVE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256679.3 5750 UTR 3 XP_005256736.1
XM_011523926.1 5750 UTR 3 XP_011522228.1
XM_011523928.2 5750 UTR 3 XP_011522230.1
XM_011523933.1 5750 UTR 3 XP_011522235.1
XM_017024733.1 5750 UTR 3 XP_016880222.1
Gene
CYB5D2
Gene Name
cytochrome b5 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details