Product Details

SNP ID
rs6698793
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:202883973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGCTAGGGCCAGGAGATTAGCTA[C/T]AACATCCGGAATGTAATGAGGCTTT
Phenotype
MIM: 614522 MIM: 603417
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL12 PubMed Links

Gene Details

Gene
KLHL12
Gene Name
kelch like family member 12
There are no transcripts associated with this gene.

Gene
LOC148709
Gene Name
actin pseudogene
There are no transcripts associated with this gene.

Gene
RABIF
Gene Name
RAB interacting factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002871.4 Intron NP_002862.2

View Full Product Details