Product Details

SNP ID
rs9818538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185296642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTGTACTTATCTCTGATTGGC[A/G]TTCTCTTCATAGGTATTTTCCCCAG
Phenotype
MIM: 604915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K13 PubMed Links

Gene Details

Gene
MAP3K13
Gene Name
mitogen-activated protein kinase kinase kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242314.1 Intron NP_001229243.1
NM_001242317.1 Intron NP_001229246.1
NM_004721.4 Intron NP_004712.1
XM_011513310.2 Intron XP_011511612.1
XM_017007456.1 Intron XP_016862945.1
XM_017007457.1 Intron XP_016862946.1
XM_017007458.1 Intron XP_016862947.1
XM_017007459.1 Intron XP_016862948.1

View Full Product Details