Product Details

SNP ID
rs9545126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:79483546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTAATAAACTGGAGCAATAAACC[A/C]CTTGAGTTGTTGAATGTTAAGGGAT
Phenotype
MIM: 610041
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NDFIP2 PubMed Links

Gene Details

Gene
NDFIP2
Gene Name
Nedd4 family interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161407.1 Intron NP_001154879.1
NM_019080.2 Intron NP_061953.2
XM_006719840.3 Intron XP_006719903.1
Gene
NDFIP2-AS1
Gene Name
NDFIP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details