Product Details

SNP ID
rs9526021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44944506 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAATCCATTATTATGATTGAATA[C/T]TGAACGTTCCAATCTCAGTAATTAA
Phenotype
MIM: 604354
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUFIP1 PubMed Links

Gene Details

Gene
NUFIP1
Gene Name
NUFIP1, FMR1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012345.2 Intron NP_036477.2

View Full Product Details