Product Details

SNP ID
rs9833853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:182807123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAGACCTTCCCAAAGCCAATAG[C/T]CAATTAGTGTCACAGCCAGGACTTG
Phenotype
MIM: 605869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP11B PubMed Links

Gene Details

Gene
ATP11B
Gene Name
ATPase phospholipid transporting 11B (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014616.2 Intron NP_055431.1
XM_005247241.2 Intron XP_005247298.1
XM_005247243.2 Intron XP_005247300.1
XM_011512593.2 Intron XP_011510895.1
XM_011512594.2 Intron XP_011510896.1
XM_011512597.2 Intron XP_011510899.1
XM_017006008.1 Intron XP_016861497.1
XM_017006009.1 Intron XP_016861498.1

View Full Product Details