Product Details

SNP ID
rs10433488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:189960075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTGGCTTTCTTTGTTTCTTGG[C/T]GACTCCAAACATCTTATTGTCTCTG
Phenotype
MIM: 610341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P3H2 PubMed Links

Gene Details

Gene
P3H2
Gene Name
prolyl 3-hydroxylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134418.1 Intron NP_001127890.1
NM_018192.3 Intron NP_060662.2
XM_011512955.1 Intron XP_011511257.1

View Full Product Details