Product Details

SNP ID
rs9905402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8003108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCCGGGCTCTGCGGTCCCGCG[C/T]GGTGGGCTCCGTCCCTGCCCCGCCT
Phenotype
MIM: 600179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCY2D PubMed Links

Gene Details

Gene
GUCY2D
Gene Name
guanylate cyclase 2D, retinal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000180.3 61 Missense Mutation CGG,TGG R21W NP_000171.1
XM_011523816.1 61 Missense Mutation CGG,TGG R21W XP_011522118.1

View Full Product Details