Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_032818.2 | 3513 | UTR 3 | NP_116207.2 | ||
XM_011518057.2 | 3513 | Intron | XP_011516359.1 | ||
XM_017015225.1 | 3513 | Intron | XP_016870714.1 | ||
XM_017015226.1 | 3513 | Intron | XP_016870715.1 | ||
XM_017015227.1 | 3513 | Intron | XP_016870716.1 | ||
XM_017015228.1 | 3513 | Intron | XP_016870717.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195200.1 | 3513 | Intron | NP_001182129.1 | ||
NM_001195201.1 | 3513 | Intron | NP_001182130.1 | ||
NM_001195217.1 | 3513 | Intron | NP_001182146.1 | ||
NM_174923.2 | 3513 | Intron | NP_777583.2 | ||
XM_005251403.4 | 3513 | Intron | XP_005251460.1 |