Product Details

SNP ID
rs10515236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:96391159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAACTTCCAAGGACAGAGTGATTC[C/T]GCAAGTCTTTTAGGAGAACATTCTC
Phenotype
MIM: 162150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929710 PubMed Links

Gene Details

Gene
LOC101929710
Gene Name
uncharacterized LOC101929710
There are no transcripts associated with this gene.

Gene
PCSK1
Gene Name
proprotein convertase subtilisin/kexin type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000439.4 4343 UTR 3 NP_000430.3
NM_001177875.1 4343 UTR 3 NP_001171346.1

View Full Product Details