Product Details

SNP ID
rs9480924
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109295867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTCCACACTGCACAGCTATCTG[C/T]TTGATTCCACATTCACGGGCTATAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC162P PubMed Links

Gene Details

Gene
CCDC162P
Gene Name
coiled-coil domain containing 162, pseudogene
There are no transcripts associated with this gene.

Gene
LOC100996634
Gene Name
transmembrane protein FLJ37396
There are no transcripts associated with this gene.

View Full Product Details