Product Details

SNP ID
rs10072253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132074075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATAACATGACATTTTCCTTTT[C/T]TACAGAATGAAACAGTAGAAGTCAT
Phenotype
MIM: 138960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSF2 PubMed Links

Gene Details

Gene
CSF2
Gene Name
colony stimulating factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000758.3 Intron NP_000749.2

View Full Product Details