Product Details

SNP ID
rs9344255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:63541980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAACTTTTTTACATAGAATTTCC[C/T]CCATGTTTTAAAAACTCTCATTTGG
Phenotype
MIM: 611470 MIM: 601585
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGSN PubMed Links

Gene Details

Gene
LGSN
Gene Name
lengsin, lens protein with glutamine synthetase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143940.1 Intron NP_001137412.1
NM_016571.2 Intron NP_057655.2
XM_011535889.2 Intron XP_011534191.1
XM_011535892.2 Intron XP_011534194.1
XM_017010929.1 Intron XP_016866418.1
XM_017010930.1 Intron XP_016866419.1
XM_017010931.1 Intron XP_016866420.1
Gene
PTP4A1
Gene Name
protein tyrosine phosphatase type IVA, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003463.4 Intron NP_003454.1
XM_011536111.1 Intron XP_011534413.1
XM_011536112.1 Intron XP_011534414.1
XM_017011270.1 Intron XP_016866759.1
XM_017011271.1 Intron XP_016866760.1
XM_017011272.1 Intron XP_016866761.1

View Full Product Details