Product Details

SNP ID
rs9456447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159780331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAATGTTATTTTATCCCATGC[C/G]TATAACTGCTCGTATCACTGTGAGA
Phenotype
MIM: 100678 MIM: 611831 MIM: 186980
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACAT2 PubMed Links

Gene Details

Gene
ACAT2
Gene Name
acetyl-CoA acetyltransferase 2
There are no transcripts associated with this gene.

Gene
MRPL18
Gene Name
mitochondrial ribosomal protein L18
There are no transcripts associated with this gene.

Gene
SNORA20
Gene Name
small nucleolar RNA, H/ACA box 20
There are no transcripts associated with this gene.

Gene
SNORA29
Gene Name
small nucleolar RNA, H/ACA box 29
There are no transcripts associated with this gene.

Gene
TCP1
Gene Name
t-complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008897.1 Intron NP_001008897.1
NM_030752.2 Intron NP_110379.2

View Full Product Details