Product Details

SNP ID
rs9930692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69791148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGGTCAAAGCAGAAGTTGGCCT[G/A]TTGGACCTGGTAACATCCTGAGGTC
Phenotype
MIM: 602308
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
WWP2 PubMed Links

Gene Details

Gene
WWP2
Gene Name
WW domain containing E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270453.1 Intron NP_001257382.1
NM_001270454.1 Intron NP_001257383.1
NM_001270455.1 Intron NP_001257384.1
NM_007014.4 Intron NP_008945.2
NM_199424.2 Intron NP_955456.1
XM_011522823.2 Intron XP_011521125.1
XM_011522825.1 Intron XP_011521127.1
XM_011522826.2 Intron XP_011521128.1
XM_017022879.1 Intron XP_016878368.1
XM_017022880.1 Intron XP_016878369.1
XM_017022881.1 Intron XP_016878370.1

View Full Product Details