Product Details

SNP ID
rs6685758
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:11280285 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGAAAGGTCTGTGTGAGCCAGAG[A/G]GGGCATGAGACACAGTTGGTGAGTT
Phenotype
MIM: 611632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBIAD1 PubMed Links

Gene Details

Gene
UBIAD1
Gene Name
UbiA prenyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013319.2 Intron NP_037451.1
XM_006710590.3 Intron XP_006710653.1
XM_011541304.2 Intron XP_011539606.1

View Full Product Details