Product Details

SNP ID
rs6026142
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58239574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATTTGCATCCACATAAAATTCA[A/G]GCCAATAAGCCTTGCTGCAGCTTCC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PPP4R1L PubMed Links

Gene Details

Gene
PPP4R1L
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details