Product Details

SNP ID
rs10416859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20413684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTAATATGCAATTTAAACAAT[G/T]CTCTTAAGGTTTTCTAGGGTAATTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF826P PubMed Links

Gene Details

Gene
ZNF826P
Gene Name
zinc finger protein 826, pseudogene
There are no transcripts associated with this gene.

View Full Product Details