Product Details

SNP ID
rs9383891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151412916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCTGATGATCCAGTCACCCCCC[A/C]ACCAGGACCCACCTCCAACACGGGC
Phenotype
MIM: 614917
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RMND1 PubMed Links

Gene Details

Gene
RMND1
Gene Name
required for meiotic nuclear division 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271937.1 Intron NP_001258866.1
NM_017909.3 Intron NP_060379.2
XM_005267040.3 Intron XP_005267097.1
XM_017010988.1 Intron XP_016866477.1

View Full Product Details