Product Details

SNP ID
rs10001093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186220788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAATTTTATTTTATTGTGATGT[C/G]CTTGTTTGGCTCTGCTATCAAAGTA
Phenotype
MIM: 608614 MIM: 229000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP4V2 PubMed Links

Gene Details

Gene
CYP4V2
Gene Name
cytochrome P450 family 4 subfamily V member 2
There are no transcripts associated with this gene.

Gene
KLKB1
Gene Name
kallikrein B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000892.4 Intron NP_000883.2
NM_001318394.1 Intron NP_001305323.1
NM_001318396.1 Intron NP_001305325.1
XM_011531930.2 Intron XP_011530232.1
XM_017008181.1 Intron XP_016863670.1
XM_017008182.1 Intron XP_016863671.1
XM_017008183.1 Intron XP_016863672.1
XM_017008184.1 Intron XP_016863673.1

View Full Product Details