Product Details

SNP ID
hCV30201632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53938060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGCTTGAACGATGACCTGTCGT[C/G]TGGAAGTGCAAAGGCCCAGGAGTGA
Phenotype
MIM: 300560
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PHF8 PubMed Links

Gene Details

Gene
PHF8
Gene Name
PHD finger protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184896.1 4678 UTR 3 NP_001171825.1
NM_001184897.1 4678 Intron NP_001171826.1
NM_001184898.1 4678 Intron NP_001171827.1
NM_015107.2 4678 UTR 3 NP_055922.1

View Full Product Details