Product Details

SNP ID
rs9935945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11549775 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAGAGAGAGACACACGGAGCGC[A/G]TTACTGATCACAACAGGGTGAACAC
Phenotype
MIM: 603795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LITAF PubMed Links

Gene Details

Gene
LITAF
Gene Name
lipopolysaccharide induced TNF factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136472.1 Intron NP_001129944.1
NM_001136473.1 Intron NP_001129945.1
NM_004862.3 Intron NP_004853.2
XM_006720982.2 Intron XP_006721045.1
XM_006720983.3 Intron XP_006721046.1
XM_006720984.3 Intron XP_006721047.1
XM_006720985.3 Intron XP_006721048.1
XM_011522754.2 Intron XP_011521056.1
XM_017023896.1 Intron XP_016879385.1

View Full Product Details