Product Details

SNP ID
rs6087528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33823138 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAACATTGTATTTCAAAGAAAAA[A/T]TTTTTTTTTTTCAAAAAAAGTATCC
Phenotype
MIM: 610897
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHMP4B PubMed Links

Gene Details

Gene
CHMP4B
Gene Name
charged multivesicular body protein 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176812.4 Intron NP_789782.1

View Full Product Details