Product Details

SNP ID
rs9876642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100261867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAGGTTGTACAACATCATTTTT[A/T]AAAAAGGGACCTTTAGAGGCTTGTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TBC1D23 PubMed Links

Gene Details

Gene
TBC1D23
Gene Name
TBC1 domain family member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199198.2 Intron NP_001186127.1
NM_018309.4 Intron NP_060779.2
XM_011512974.2 Intron XP_011511276.1
XM_017006840.1 Intron XP_016862329.1
XM_017006841.1 Intron XP_016862330.1

View Full Product Details