Product Details

SNP ID
rs9658936
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:150266588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCCAGCTGTGATGAAGGCCCA[A/G]AGCCCCATGGAAACAGTGACTGCAT
Phenotype
MIM: 607629 MIM: 604832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APH1A PubMed Links

Gene Details

Gene
APH1A
Gene Name
aph-1 homolog A, gamma-secretase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077628.2 1077 Silent Mutation CTC,CTT L226L NP_001071096.1
NM_001243771.1 1077 Silent Mutation CTC,CTT L169L NP_001230700.1
NM_001243772.1 1077 Silent Mutation CTC,CTT L156L NP_001230701.1
NM_016022.3 1077 Silent Mutation CTC,CTT L226L NP_057106.2
XM_017001417.1 1077 Silent Mutation CTC,CTT L169L XP_016856906.1
Gene
C1orf54
Gene Name
chromosome 1 open reading frame 54
There are no transcripts associated with this gene.

Gene
CA14
Gene Name
carbonic anhydrase 14
There are no transcripts associated with this gene.

View Full Product Details