Product Details

SNP ID
rs9320345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:96905020 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTGTATCTGTCCCTCCATGTCC[C/T]CATGAACTTGCTTTATATGGAAGTA
Phenotype
MIM: 611776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL32 PubMed Links

Gene Details

Gene
KLHL32
Gene Name
kelch like family member 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286250.1 Intron NP_001273179.1
NM_001286251.1 Intron NP_001273180.1
NM_001286252.1 Intron NP_001273181.1
NM_001286254.2 Intron NP_001273183.1
NM_001323252.1 Intron NP_001310181.1
NM_001323253.1 Intron NP_001310182.1
NM_001323254.1 Intron NP_001310183.1
NM_001323255.1 Intron NP_001310184.1
NM_001323256.1 Intron NP_001310185.1
NM_001323257.1 Intron NP_001310186.1
NM_001323258.1 Intron NP_001310187.1
NM_001323259.1 Intron NP_001310188.1
NM_001323260.1 Intron NP_001310189.1
NM_001323261.1 Intron NP_001310190.1
NM_001323262.1 Intron NP_001310191.1
NM_001323263.1 Intron NP_001310192.1
NM_001323264.1 Intron NP_001310193.1
NM_052904.3 Intron NP_443136.2
XM_005266813.4 Intron XP_005266870.1
XM_017010228.1 Intron XP_016865717.1
XM_017010229.1 Intron XP_016865718.1
Gene
NDUFAF4
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 4
There are no transcripts associated with this gene.

View Full Product Details