Product Details
- SNP ID
-
rs6119733
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31846922 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATGCAGCACCCTCATCAAGAGC[C/T]TAGTATGGAGTAGGGATATGAACAT
- Phenotype
-
MIM: 616776
MIM: 602939
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DUSP15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76141115] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DUSP15
- Gene Name
- dual specificity phosphatase 15
- Gene
- FOXS1
- Gene Name
- forkhead box S1
There are no transcripts associated with this gene.
View Full Product Details