Product Details

SNP ID
rs9587153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:106547870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGCCTAATTAAAAAAACAAGAT[A/T]TTTAAAGTAAAGCAACTTCCTTCAG
Phenotype
MIM: 614046
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARGLU1 PubMed Links

Gene Details

Gene
ARGLU1
Gene Name
arginine and glutamate rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018011.3 Intron NP_060481.3

View Full Product Details