Product Details
- SNP ID
-
rs7527285
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:248838356 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGACTGACCACTAGCCGACGCGGGC[A/G]AGAGGGACAGGAGCGTGACCTCCCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF672
PubMed Links
Gene Details
- Gene
- ZNF672
- Gene Name
- zinc finger protein 672
View Full Product Details