Product Details

SNP ID
rs8176586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:187469171 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAATAAGGACTAGAGTGTCTTG[A/G]TATAGTAACTCAGGTAATAAAAAAT
Phenotype
MIM: 152310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFPI PubMed Links

Gene Details

Gene
TFPI
Gene Name
tissue factor pathway inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005246818.1 Intron XP_005246875.1
XM_005246819.1 Intron XP_005246876.1
XM_006712720.3 Intron XP_006712783.1
XM_011511707.2 Intron XP_011510009.1
XM_011511708.1 Intron XP_011510010.1
XM_011511709.2 Intron XP_011510011.1

View Full Product Details