Product Details

SNP ID
rs6012020
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46720649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGTTGGGTCATGGCGTGCTTTC[C/T]TGATTCCCACAAACTGACCCTTGTG
Phenotype
MIM: 606145
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SLC2A10 PubMed Links

Gene Details

Gene
SLC2A10
Gene Name
solute carrier family 2 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030777.3 Intron NP_110404.1

View Full Product Details