Product Details

SNP ID
rs10399812
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:42432537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAGATTTTCAATAAATGTTTGAA[G/T]GAATGAGAGAATCAACCAATAAGTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RIMKLA PubMed Links

Gene Details

Gene
RIMKLA
Gene Name
ribosomal modification protein rimK like family member A
There are no transcripts associated with this gene.

Gene
ZMYND12
Gene Name
zinc finger MYND-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146192.1 Intron NP_001139664.1
NM_032257.4 Intron NP_115633.3

View Full Product Details