Product Details

SNP ID
rs10420381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20126481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCATCGCGTGCCTAGGCAAAACT[C/T]CATTTCAAAAACAAAACAAAAAAAT
Phenotype
MIM: 603973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105372310 PubMed Links

Gene Details

Gene
LOC105372310
Gene Name
uncharacterized LOC105372310
There are no transcripts associated with this gene.

Gene
ZNF90
Gene Name
zinc finger protein 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007138.1 Intron NP_009069.1

View Full Product Details