Product Details
- SNP ID
-
rs3211097
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:227329764 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATTTAAATGAGTAAAGGAACAAGC[A/T]GTGACACATCACTAGGAAGGTCAGT
- Phenotype
-
MIM: 614785
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC654841
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3211098] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC654841
- Gene Name
- uncharacterized LOC654841
There are no transcripts associated with this gene.
- Gene
- MFF
- Gene Name
- mitochondrial fission factor
View Full Product Details