Product Details

SNP ID
rs6567569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:3606268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATGATCATCCATACGGACATTC[C/T]GCACCGTGATTGTTTCGTGATCATC
Phenotype
MIM: 300083
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKX PubMed Links

Gene Details

Gene
PRKX
Gene Name
protein kinase, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005044.4 Intron NP_005035.1

View Full Product Details