Product Details

SNP ID
rs7532507
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:244659474 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACAGAATTCAGTTGCTTATATT[G/T]GTAGGTCTGAGGTCCCATTCCTTCC
Phenotype
MIM: 614638
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DESI2 PubMed Links

Gene Details

Gene
DESI2
Gene Name
desumoylating isopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297746.1 Intron NP_001284675.1
NM_016076.4 Intron NP_057160.2
XM_011544203.2 Intron XP_011542505.1
XM_011544205.2 Intron XP_011542507.1
XM_011544206.2 Intron XP_011542508.1
XM_011544207.2 Intron XP_011542509.1

View Full Product Details