Product Details

SNP ID
rs8129374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33269509 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTACATCAGAGAAGAAGTAGAAAG[A/G]TGAATAAGATAGTCCTCGTCTGTGC
Phenotype
MIM: 602376 MIM: 123889
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNAR2 PubMed Links

Gene Details

Gene
IFNAR2
Gene Name
interferon alpha and beta receptor subunit 2
There are no transcripts associated with this gene.

Gene
IL10RB
Gene Name
interleukin 10 receptor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000628.4 Intron NP_000619.3
Gene
IL10RB-AS1
Gene Name
IL10RB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details