Product Details

SNP ID
rs7514099
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:40261607 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTAGTTCCTTAAAGAAATAGTTA[A/G]GTCTTGTTCATCATTTTATACCTAG
Phenotype
MIM: 606480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMCO2 PubMed Links

Gene Details

Gene
TMCO2
Gene Name
transmembrane and coiled-coil domains 2
There are no transcripts associated with this gene.

Gene
ZMPSTE24
Gene Name
zinc metallopeptidase STE24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005857.4 Intron NP_005848.2

View Full Product Details