Product Details

SNP ID
rs5748131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19332240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAACTTTAACACTGGACGAGAAGG[C/T]CTAAGATCTTGAATTAACGGTGACA
Phenotype
MIM: 600237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIRA PubMed Links

Gene Details

Gene
HIRA
Gene Name
histone cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003325.3 Intron NP_003316.3

View Full Product Details