Product Details

SNP ID
rs9430611
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:9654189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAAACAGACTACATTGGCCAC[A/G]CTACTCACTTAGATGTTTCTGGAAT
Phenotype
MIM: 602839
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3CD PubMed Links

Gene Details

Gene
PIK3CD
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005026.3 Intron NP_005017.3
XM_006710687.2 Intron XP_006710750.1
XM_006710689.2 Intron XP_006710752.1
XM_017001476.1 Intron XP_016856965.1
XM_017001477.1 Intron XP_016856966.1
XM_017001478.1 Intron XP_016856967.1
XM_017001479.1 Intron XP_016856968.1
XM_017001480.1 Intron XP_016856969.1
XM_017001481.1 Intron XP_016856970.1
XM_017001482.1 Intron XP_016856971.1
XM_017001483.1 Intron XP_016856972.1
XM_017001484.1 Intron XP_016856973.1
XM_017001485.1 Intron XP_016856974.1
Gene
PIK3CD-AS1
Gene Name
PIK3CD antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details