Product Details

SNP ID
rs4829575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135353604 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACTATGGTCTCAAAACCAGGAGA[C/G]ATGATTAAATTTAAGGTAACAGGCT
Phenotype
MIM: 300627 MIM: 314997
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF449 PubMed Links

Gene Details

Gene
ZNF449
Gene Name
zinc finger protein 449
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152695.5 Intron NP_689908.3
XM_011531312.2 Intron XP_011529614.1
XM_011531313.2 Intron XP_011529615.1
XM_011531314.2 Intron XP_011529616.1
XM_017029350.1 Intron XP_016884839.1
XM_017029351.1 Intron XP_016884840.1
XM_017029352.1 Intron XP_016884841.1
Gene
ZNF75D
Gene Name
zinc finger protein 75D
There are no transcripts associated with this gene.

View Full Product Details