Product Details

SNP ID
rs6413529
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:153959794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCAGCCTCCCATGTGAGCCTGT[C/T]CTTATGTATAGTGTCCAACCTCTGA
Phenotype
MIM: 607138 MIM: 608972 MIM: 604740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
There are no transcripts associated with this gene.

Gene
CRTC2
Gene Name
CREB regulated transcription coactivator 2
There are no transcripts associated with this gene.

Gene
MIR6737
Gene Name
microRNA 6737
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271957.1 1627 UTR 3 NP_001258886.1
NM_001271958.1 1627 UTR 3 NP_001258887.1
NM_001271959.1 1627 UTR 3 NP_001258888.1
NM_001271960.1 1627 UTR 3 NP_001258889.1
NM_001271961.1 1627 UTR 3 NP_001258890.1
NM_014437.4 1627 UTR 3 NP_055252.2

View Full Product Details